Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BEEM™ Flat Embedding Molds, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
BEEM™
70904-12 70904-01
100489-192PK 186.94 USD InStock
100489-192 100493-032
BEEM™ Flat Embedding Molds, Electron Microscopy Sciences
Moulds Moulds for Histology
The cavity measures L x W x D: 12 x 5 x 3 mm

Molded from polyethylene. The critical surfaces of the mold are transparent, facilitating accurate specimen orientation through bottom illumination. Molds are reusable and each mold is supplied with a protective box.

The cavity measures L x W x D: 12 x 5 x 3 mm
Overall mold measures L x W x D: 52 x 40 x 5 mm
Storage box measures L x W x D: 57 x 45 x 12.5 mm (2 1/3 x 1 3/4 x 1/2")

*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc

*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty