Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70952-10 70952-05 70952-25 70951-05 70951-02 70951-10 70951-01 70952-01 70952-02 70952-50
100489-198PK 195 USD InStock
100489-198 100489-196 100489-194 100489-208 100489-206 100501-216 100489-204 100501-214 100489-202 100489-200
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Pipettes Graduated Pipettes
Made from translucent polypropylene.
  • Resistant to breakage
  • Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty