Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Opto-Wipes™ Precision Lens Cleaning Wipes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71755-03 71755-01 LCW-304 71755-02 71755-04
100489-362PK 84.26 USD InStock
100489-362 100489-360 100489-358 76248-124 100489-364
Opto-Wipes™ Precision Lens Cleaning Wipes, Electron Microscopy Sciences
Wipes Disposable Wipes and Towels Lens Papers
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments.
  • This special material offers softness, is lint-free, and traps particles and residue quite easily
Wipes made of super-soft fibers prevent the scratching of precision surfaces and its strong fiber matrix maintains integrity when wet. Opto-Wipes™ are highly absorbent which retains and withstands solvents such as MEK, methylene chloride, acetone, and alcohol. Its efficient particle trapping matrix lifts particles and residue away from the surface. They are washable and reusable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty