Would you like to log in again?
Single Edge Razor Blade for Clean Rooms, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71952-01 71952-10
100489-418PK 93.65 USD InStock
100489-418 100493-554
Single Edge Razor Blade for Clean Rooms, Electron Microscopy Sciences
Razors and Blades
These blades and dispenser springs are degreased in order to minimize particulates and residual grinding oils on the blades.
The blades are made from surgical stainless steel (significantly more durable then carbon steel) with a steel back that produces less particulate than the aluminum back type. Packed 10 blades per dispenser, 200 dispensers per case. The blade including steel back measures 39 x 19 x 0.23 mm (1 1/2 x 3/4 x 0.009").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty