Would you like to log in again?
Caps for Syringes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72500-05 72500-10
100502-740PK 87.46 USD InStock
100502-740 100489-464
Caps for Syringes, Electron Microscopy Sciences
Closures Bottle Closures
One size fits-all syringe caps that can be used with either a luer slip or luer locking syringe tip
These tips make the syringes suitable for usage as embedding media storage vessels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty