Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Polyethylene Vials; Poly-Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72621-92 72621-20 72621-62 72621-22 72621-42 72621-38 72621-40 72621-10 72621-32 72621-90 72621-30 72621-12 72621-36 72621-60
100500-200CS 430.52 USD InStock
100500-200 100489-472 100501-906 100489-482 100489-480 100501-734 100500-192 100489-478 100500-194 100500-196 100489-476 100489-474 100500-198 100500-190
Polyethylene Vials; Poly-Vials, Electron Microscopy Sciences
Vials
These flat bottom cylindrical vials have a a friction-fit, flip-top closure which can not be lost. Ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
  • Unbreakable
  • Withstand temps up to 90 °C
  • Wall thickness 1 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty