Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tooled Neck Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72646-07 72646-03 72646-02 72646-05 72646-01 72646-04
100489-528PK 214.44 USD InStock
100489-528 100489-526 100489-524 100489-522 100489-532 100489-530
Tooled Neck Vials, Electron Microscopy Sciences
Vials
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty