Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PRO-LINE Archival Sleeving - Digital Output Sleeving, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
73921-CO 73915-CO 73922-CS 73923-CS 73920-CO 73919-CO 73917-CO 73921-CS 73917-CS 73920-CS 73918-CS 73922-CO 73915-CS 73919-CS 73918-CO 73916-CO 73916-CS 73923-CO
100489-572PK 154.26 USD InStock
100489-572 100489-582 100489-560 100489-570 100489-580 100489-558 100489-568 100489-578 100489-556 100489-566 100489-576 100489-554 100489-586 100489-564 100489-574 100489-562 100489-584 100502-748
PRO-LINE Archival Sleeving - Digital Output Sleeving, Electron Microscopy Sciences
Film Processing Supplies
PRO-LINE Archival Sleeving is a specially formulated 3-mil archival polypropylene which has a physical density that is less than other sleeving materials, making it more pliable and gentle to the film
Strong and durable, PRO-LINE won’t become brittle or abrade film. PRO-LINE sleeving is optically clear, neutral in color, and has a unique anti-Newton ring surface that reduces distracting Newton ring as well as minimizing the actual surface area that is in contact with your film. All sleeving is available in clear both sides and clear front with frosted back for even light distribution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty