Would you like to log in again?
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74006-50 74008-10 74006-10 74008-50
100489-594PK 67.22 USD InStock
100489-594 100489-592 100489-598 100489-596
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
Envelopes
C-Shell™ is designed to provide filling, storage, and protection for compact disks or DVD’s during shipping
Paper Sleeves are economical per sleeve and are far less than plastic jewel case. They are space saving, durable and have clarity.
The C-Shell™ is an ultra slim (only 4.5 mm thick), one-piece polypropylene shell with a living hinge.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty