Would you like to log in again?
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64254 64260 64257-30 64258 64251-22 64256 64250 64252
100489-758PK 334.98 USD InStock
100489-758 100489-756 100489-766 100489-764 102095-026 100489-762 100489-760 100488-918
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Bottles Glass Bottles
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty