Would you like to log in again?
Polypropylene Film Sleeves, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
73939 73931 73901 73930 73948 73900 73938 73940 73944 73902 73946 73937 73935 73934 73936 73932 73933 73941
100489-910PK 42.9 USD InStock
100489-910 100489-888 100489-920 100489-886 100489-930 100489-884 100489-918 100489-928 100496-352 100489-916 100489-926 100496-350 100489-914 100489-924 100489-912 100489-922 100496-348 100501-258
Polypropylene Film Sleeves, Electron Microscopy Sciences
Film Processing Supplies
Polyguard Polypropylene film sleeves are the finest archival quality material, and are guaranteed to give you ultimate protection for your most important film negatives. Polyguard is 100% acid free, as well as being free from optical distortion. It is resistant to water, grease, oil, alkalies, and organic solvents. Polyguard is durable, yet pliable and gentle to the film and will not become brittle, crack, or abrade the film. All sizes are available in a clear open flap, clear front with frosted backing, and sealed flap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty