Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CXCR6 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-276-100
10049-316EA 514 USD InStock
10049-316
Anti-CXCR6 Rabbit Polyclonal Antibody
Antibodies
CXCR6 (Bonzo), an orphan G protein coupled receptor, has been identified as a co-receptor involved in the infection of cells by the human immunodeficiency virus (HIV) and simian immunodeficiency virus (SIV).

Type: Primary
Antigen: CXCR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty