Would you like to log in again?
HighDef® Blue IHC Chromogen (AP), Enzo Life Sciences
Supplier: Enzo Life Sciences
950-150-0030
10049-336EA 236 USD InStock
10049-336
HighDef® Blue IHC Chromogen (AP), Enzo Life Sciences
Stains Microbiology Stains
High definition IHC chromogen producing brilliant blue color for AP-activated development.
HighDef® blue IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems.
Ordering information: Contains HighDef® blue IHC chromogen (AP) substrate buffer and chromogen, Empty mixing bottle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty