Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APP Mouse Monoclonal Antibody [clone: 3E9]

Supplier: Enzo Life Sciences
ADI-NBA-100-E
10049-416EA 452 USD InStock
10049-416
Anti-APP Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Deposition of APP (Amyloid A4 protein) precursor intraneuronally as neurofibrillary tangles, extracellularly as plaques, and in blood vessels, is characteristic of both Alzheimers disease and aged Downs Syndrome. There are five forms of APP: APP395, APP563, APP695, APP751, and APP770. They are produced by alternative splicing of the APP gene. APP695 is the most predominant form in neuronal tissue while APP751 is the predominant form found elsewhere.

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty