Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Mouse Monoclonal Antibody [clone: 10000]

Supplier: Enzo Life Sciences
ADI-SAB-500-E
10049-462EA 406 USD InStock
10049-462
Anti-GFP Mouse Monoclonal Antibody [clone: 10000]
Antibodies
The green fluorescent protein (GFP) is derived from the jellyfish Aequorea victoria, that fluoresces green when exposed to blue light. GFP is a frequently used reporter for gene expression in recombinant DNA and cell transfection systems.

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Species independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty