Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cu/Zn SOD Rabbit Polyclonal Antibody (Biotin)

Supplier: Enzo Life Sciences
SOD-100B-0050
10049-480EA 294 USD InStock
10049-480
Anti-Cu/Zn SOD Rabbit Polyclonal Antibody (Biotin)
Antibodies
Superoxide dismutase (SOD) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells: extracellular SOD (EC SOD, SOD-3); copper and zinc-containing SOD (Cu/Zn SOD, SOD-1); and manganese-containing SOD (Mn SOD, SOD-2). The Cu/Zn form binds Cu and Zn ions and exists as a soluble 32kDa homodimer in the cytosol. Mutations in the SOD-1 gene accounts for approximately 20% of amyelotrophic lateral sclerosis (ALS) or ‘Lou Gehrig disease,’ a neurodegenerative disorder characterized by the death of motor neurons in the brain, brainstem, and spinal cord that results in fatal paralysis.

Type: Primary
Antigen: Cu/Zn SOD
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty