Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40 Rat Monoclonal Antibody [clone: FGK45]

Supplier: Enzo Life Sciences
805-046PF-C500
10049-628EA 663 USD InStock
10049-628
Anti-CD40 Rat Monoclonal Antibody [clone: FGK45]
Antibodies
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: FGK45
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty