Would you like to log in again?
LBP, soluble (mouse) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-305-KI01
10049-636EA 1140 USD InStock
10049-636
LBP, soluble (mouse) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of mouse and rat soluble as well as bound LBP levels in serum, plasma and cell culture medium.
Ordering information: Contains 1 precoated ELISA module1 vial anti-LBP (mouse) detecting MAb1 vial LBP standard (mouse)1 vial reference serum (mouse)2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution1 vial substrate solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty