Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDC25A Mouse Monoclonal Antibody [clone: DCS-120]

Supplier: Enzo Life Sciences
KAM-CC085-E
10049-866EA 358 USD InStock
10049-866
Anti-CDC25A Mouse Monoclonal Antibody [clone: DCS-120]
Antibodies
The cell division cycle (cdc) group of proteins consists of many protein classes, including cyclin dependent kinases (Cdks), phosphoprotein phosphatases, mitogen activated kinases, small GTPases, and cyclins. These proteins play a central role in regulating progression through the cell cycle through the formation and/or modification of key substrates. Cdc42 belongs to the Rho-family of GTPases, and promotes actin polymerization to form filopodia. Cdc25A is a dual-specificity phosphatase that acts on phosphorylated threonine and tyrosine residues of Cdks, leading to binding and activation of their partner cyclins.

Type: Primary
Antigen: CDC25A
Clonality: Monoclonal
Clone: DCS-120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty