Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-ST210-E
10049-874EA 442 USD InStock
10049-874
Anti-S Rabbit Polyclonal Antibody
Antibodies
Protein phosphorylation is a complex biochemical process that is regulated by protein kinases that catalyze the transfer of the gamma-P of ATP to a recipient protein that acts as a substrate. This post-translational modification is primarily directed onto serine, threonine, and tyrosine amino acid residues within a protein. Changes in the phosphorylation status of phosphoproteins can alter their biological function, and aberrations in signal transduction cascades have been linked to a number of diseases, including cancer, diabetes, heart disease, inflammation and neurological disorders.

Type: Primary
Antigen: S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty