Would you like to log in again?
Cryo-Sleeve, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61082-10
100490-098PK 222.52 USD InStock
100490-098
Cryo-Sleeve, Electron Microscopy Sciences
Cryogenic Canes
Clear polyvinyl chloride. Ideal for enclosing Cryo Storage Canes for extra security during handling and storage. Fits over most other canes. Will not become brittle during freezing or thawing. Will not shred or tear after repeated use like cardboard. Length: 27.5 cm (10 13/16").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty