Would you like to log in again?
Emery Paper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62117-10 62117-30 62117-03 62117-04 62117-20 62117-01
100490-132PK 67.22 USD InStock
100490-132 100490-134 100490-140 100490-128 100490-136 100490-138
Emery Paper, Electron Microscopy Sciences
Abrasives
This emery paper comes in sheets and in varying grit sizes.
Grit 4/0 is the finest and 3 is the coarsest.
Size: 22×34 cm (9×13¾")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty