Would you like to log in again?
Biopsy Foam Pads, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
62325-01 62325-06 62325-04
100490-164PK 64.08 USD InStock
100490-164 100490-166 100490-168
Biopsy Foam Pads, Electron Microscopy Sciences
Biopsy Pads
A cellular polyester blue foam that has uniform air pockets throughout the pad for optimal fluid flow
It prevents either small or fragmented tissue specimens from being lost. To Use: Place small samples or specimen fragments between the two pads and place the specimen in a cassette or capsule.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty