Would you like to log in again?
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
63035-10 63036-PS 63034-05 63037-PP 63035-05 63035-50 63034-50 63037-PS 63036-PP 63034-10
100502-616CS 294.38 USD InStock
100502-616 100500-420 100490-274 100490-276 100500-422 100500-424 100500-416 100490-280 100500-418 100490-278
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Tubes Test Tubes
Transport vials/tubes are ideal for sample collection and storage.
  • Conical tubes with freestanding skirts
  • Tubes are autoclavable
  • Write-on area on side
Transport Vials are made from virgin polypropylene. They are available in three sizes, and come either sterile or non sterile. Graduations provide convenient and accurate references.
Transport Tubes are made from virgin polypropylene (PP) or polystyrene (PS). They also work well in Olympus automated testing equipment. Plug caps are required.
Delivery information: 5 ml and 10 ml vials are supplied with white screw cap and 50 mL vials are supplied with blue cap that has a molded-in sealing ring for a leak-proof fit. Plug caps are available in yellow color and you can choose from hollow top (HT) or hollow bottom (HB).
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty