Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tissue File Cases, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63286-01 63286-10 63286-06
100500-430CS 1406.87 USD InStock
100500-430 100490-290 100498-382
Tissue File Cases, Electron Microscopy Sciences
Boxes Storage Boxes
Tissue file case is a histological embedment block cataloging and storage system. This file system incorporates the necessary features offering embedment block protection, simplified cataloging, reduction of filing error and facilitation of retrieval. All of this while conserving space and being very affordable.
  • Embedment block capacity: 100 to 1000 per tissue file
  • Compartments: 100 of 38×25×57 mm (1½×1×2¼")
  • Compartment arrangement: 10 across×10 deep
For cataloging, each compartment is numerically indexed on a permanent chart inside the lid which also provides space for detailed information.
Tissue files are made from solid fiberboard with a protective, attractive, scuff-resistant exterior green covering. Its hinged lid lifts open and with a cloth stay support the lid can be kept in the open position.
By removing alternating partitions, each compartment can be doubled or quadrupled in size. For other storage needs, the partitions may also be removed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty