Would you like to log in again?
Narrow Mouth Ground Glass Stoppered Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64204-A 64204-C 64206-C 64207-C 64209-C 64202-A 64202-C
100498-646EA 465.84 USD InStock
100498-646 100502-018 100490-408 100490-416 100502-534 100490-410 100490-412
Narrow Mouth Ground Glass Stoppered Bottles, Electron Microscopy Sciences
Bottles Glass Bottles
New designed ground glass stoppered bottles feature a no-drip pour lid for clean pouring of contents.
The most preferred container for storage of prepared chemicals, and stock solutions. Octagon-shaped ground glass stopper is included. The borosilicate '400' glass offers a high degree of protection from leaching and the amber 500' glass  gives extra protection to light-sensitive contents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty