Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Disposable Sterile Collecting Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64244-02 64244-10 64244-01 64244-05
100490-494PK 201.94 USD InStock
100490-494 100490-496 100490-498 100490-500
Disposable Sterile Collecting Bottles, Electron Microscopy Sciences
Bottles Cell Culture Bottles
Disposable polystyrene bottles for the storage of media, buffers and other aqueous solutions.
  • Easy grip sides facilitate handling
  • Sterile
  • Certified non-pyrogenic
  • Neck size 45 mm
Plug seal cap provides an airtight seal and helps minimize the risk of contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty