Would you like to log in again?
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64298-04 64298-08 64298-02
100490-550PK 68.94 USD InStock
100490-550 100490-552 100488-920
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Bottles Dropping and Dispensing Bottles
This low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette. The pipettes are graduated for nominal indication of amount of liquid being drawn. Bellows-shaped bulb makes for easy use and provides excellent drop information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty