Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Carbon Rods, Pointed and Presharpened, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70220-45 70221-60 70220-02 70221-01 70220-01 70221-40 70220-50 70221-02
102096-590PK 46.11 USD InStock
102096-590 102096-584 100490-748 100490-744 102096-588 100490-746 102096-586 100490-742
Carbon Rods, Pointed and Presharpened, Electron Microscopy Sciences
Solid Rods
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
  • Rod Length: 60.3 mm (23/8”)
  • Neck Length: 5.2 mm (0.205”)
  • Neck Diameter: 1.14 mm (0.045”)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty