Would you like to log in again?
Vertical Staining Jar w/Glass Lid, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70318-04
100490-752PK 349.59 USD InStock
100490-752
Vertical Staining Jar w/Glass Lid, Electron Microscopy Sciences
Staining Boxes
An all glass rectangular staining jar which accommodates 8 to 16 slides and comes with a glass lid. Its wide top eliminates the need for forceps in the handling of the slides. It accommodates slides up to 103 x 39 mm (4 x 1 1/2") in size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty