Would you like to log in again?
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70326-32 70326-42 70326-30 70326-52 70326-46 70326-22 70326-16 70326-56 70326-12 70326-20 70326-10 70326-14 70326-54 70326-24 70326-40 70326-28 70326-50 70326-34 70326-36 70326-44 70326-26
100490-770PK 309.94 USD InStock
100490-770 100490-792 100490-780 100490-794 100490-772 100490-782 100490-790 100502-780 100502-782 100490-778 100490-766 100490-788 100501-302 100490-768 100490-774 100490-784 100490-762 100501-300 100490-776 100490-764 100490-786
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
Specimen Mounting Supplies
CoverWell™ Perfusion Chambers allows up to 16 different specimens on one coverglass.
  • Chambers are removable for permanent mounting of specimens
  • CoverWell™ chambers are made from medical grade silicone bonded to clear, UV transparent plastic
  • Available in multiple designs allowing up to sixteen different specimens or analytical test on one coverglass
  • Ideal for cell imaging on upright or inverted microscopes
"Press-to-Seal" covers form water-tight, multiwall cell incubation or cytochemistry chamber when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports (1.5 mm diameter) without disturbing or cross-contaminating specimens in adjacent wells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty