Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70327-13 70327-05 70327-28 70327-25 70327-04 70327-08 70327-23 70327-20 70327-10
100490-804PK 137.3 USD InStock
100490-804 100488-338 100490-800 100501-876 100490-802 100488-340 100490-796 100490-798 100488-342
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
Specimen Mounting Supplies
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty