Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HybriWell™ Sealing Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70328-17 70328-14 70328-00 70328-19 70328-13 70328-08 70328-10 70328-30 70328-12 70328-09 70328-35 70328-15 70328-01 70328-03 70328-05
100490-808PK 411.02 USD InStock
100490-808 102096-618 100490-818 102096-628 102096-616 100490-814 100490-806 100490-816 102096-622 100490-810 102096-620 100490-812 102096-626 102096-624 100490-820
HybriWell™ Sealing Chambers, Electron Microscopy Sciences
Slides Microarray Slides
HybriWell Sealing Chambers System seals securely to a microscope slide surface in seconds – for most protein and nucleic acid assays.
  • Enclose a single or multiple specimens in a small reagent volume
  • Access ports in the chamber surface allow for the addition or removal of reactants
  • Ports are easily sealed using Adhesive Seal Tabs (provided) to prevent reagent evaporation
  • Sealed chambers are watertight and ideally suited for water-bath incubations
  • RNase free, hydrophobic surfaces will not trap or bind probes
  • Disposable chamber remove cleanly and easily even after heating
  • Come with Non-Adhesive Peeling Tab
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty