Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Hybrislip™ Hybridization Cover - Rnase free, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70329-60 70329-30C 70329-62 70329-40 70329-40C 70329-25 70329-60C 70329-62C 70329-30 70329-24C 70329-22C 70329-22 70329-24 70329-45 70329-25C
102096-650PK 72.66 USD InStock
102096-650 102096-660 100490-826 100490-828 102096-644 100490-822 100490-832 102096-654 100490-824 102096-652 100490-834 102096-648 102096-658 102096-646 100490-830
Hybrislip™ Hybridization Cover - Rnase free, Electron Microscopy Sciences
Cover Glasses
HybriSlips™ are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution.
  • RNase and DNase free
  • Hydrophobic
  • Disposable
  • Will not chip or break
  • Clear – stay flat
Hybrislips™ - ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. Hybrislips™ remain flat and will not curl, even at high temperatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty