Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Press to Seal Silicone Sheet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70338-52 70338-72 70338-74 70338-40 70338-79 70338-70 70338-44 70338-29 70338-31 70338-42 70338-32 70338-05 70338-30 70338-76 70338-46 70338-10 70338-25 70338-20
100490-890PK 508.65 USD InStock
100490-890 100490-892 102681-188 100490-906 102681-190 100490-908 100490-902 100490-904 102681-196 100490-888 100490-910 100490-898 100490-900 102681-186 102681-192 100490-894 102681-194 100490-896
Press to Seal Silicone Sheet, Electron Microscopy Sciences
Slides Chamber Slides
Press-To-Seal silicone sheets allow researchers to prepare isolators for their own design chambers
The sillicone sheet material is available with 3 different gasket types either:
Silicone (S) / Silicone (S) – press-to-seal silicone gasket only, no adhesive
Silicone (S) / Adhesive (A) – press-to-seal silicone on one side and adhesive on the other side.
Adhesive (A) / Adhesive (A) – press-to-seal adhesive on both sides.
Normally silicone isolators are used to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty