Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CultureWell™ Accessories, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70466-20 70466-40 70465-2R2 70466-10 70466-30 70465-3S 70465-4R 70465-8R 70465-50R 70465-2R1
100489-176PK 114.44 USD InStock
100489-176 100489-174 100489-172 100489-182 100489-170 100489-180 100489-168 100489-178 100490-940 100501-892
CultureWell™ Accessories, Electron Microscopy Sciences
Cover Glasses
Reusable CultureWell™ gaskets from CultureWell™ Chambered Coverglass and CultureWell™ Chamber Coverglass Inserts are available bulk packaged.
  • Gaskets are ideal for forming wells on glass microscope slides or in polystyrene dishes
  • Gaskets are non-sterile and may be sterilized by autoclaving, UV or alcohol
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty