Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70950-15 70950-52 70950-55 70950-12
100490-958PK 84.41 USD InStock
100490-958 100490-956 100490-960 100490-962
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Pipettes Pasteur/Transfer Pipettes
Good for handling infectious materials, inoculating culture media and routine sampling. Available in either Borosilicate or Flint glass, with an outside body diameter of 7 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty