Would you like to log in again?
DrainRack™ Slide Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71446-W 71446-G 71446-P 71446-Y 71446-B
100490-990PK 73.14 USD InStock
100490-990 100490-992 100490-986 100490-988 100490-994
DrainRack™ Slide Rack, Electron Microscopy Sciences
Slide Holders
Made from high impact polystyrene. This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individual numbered slots.
All sides are stored upright for easy insertion and removal. A unique feature with the DrainRack™ is the ability to read bar codes without having to remove the slides from the rack. For space saving purposes, you can double the amount of slides simply by storing 2 slides per slot, giving you a capacity of 200 slides per tray instead of 100. The racks are not autoclavable. They measure: 75 x 231 x 25 mm (3 x 9 x 1").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty