Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Feather® Sterile Micro Scalpels; Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72046-30 72045-15 72046-45 72046-15 72045-30 72045-45
101413-480PK 83.11 USD InStock
101413-480 102097-826 100491-034 101413-484 100491-038 100491-036
Feather® Sterile Micro Scalpels; Stainless Steel, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
Micro precision, ultra sharp points.
  • Sterilized by gamma irradiation
  • Disposable
  • Sure-grip aluminum handle-color coded for easy angle identification
  • Comes in a carbonate plastic case
  • Available in 3 angle styles: 15°, 30° and 45°
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty