Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Two Part Conductive Silver Paint, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12642-14
100491-332KT 197.33 USD InStock
100491-332
Two Part Conductive Silver Paint, Electron Microscopy Sciences
Stains Microscopy and staining reagents
This electrically conductive silver epoxy is used for adhering samples, as well as many other purposes.
  • Electrically conductive
  • Useful in many applications
  • Ideal for bonding heat sensitive components
In addition to adhering samples, uses include solderless connections such as bonding in electric design, prototype and repair work, circuit board repair, surface mount connections, static discharge, shielding and grounding. It is also ideal for the bonding of heat sensitive components. Its curing time is within ten minutes at 100°F (38°C), or at room temperature. Conductivity is 0.001 ohm/cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty