Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silica, colloidal

Supplier: Electron Microscopy Sciences
Warning

50366-05 50366-06
100491-406GL 221.16 USD InStock
100491-406 100488-226
Silica, colloidal
Silica (amorphous)
This non-crystallizing Colloidal Silica is made to be user friendly. It eliminates the problems that are caused by drying or freezing that are associated with other colloidal silica products which are used for chemical/mechanical polishing

Colloidal silica is a first choice silica dispersion for optimizing polishing results such as silicon, fused quartz, fused silica, lithium niobate, YAZ, GGG, alexandrite, sapphire and many others.

Application: Colloidal silica can be used full strength or diluted. Dilution to 1:1 or higher with deionized water is recommended. Polyurethane and short nap cloths can be used.


Formula: SiO₂
MW: 60.083 g/mol
CAS Number: 112926-00-8
Order Now
Specification Test Results
Total Solids 29-31%
Specific Gravity 1.2
pH 9-11
Particle Size ~80 nm
Non-Drying Formula
Not for use above 140° F (60° C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty