Would you like to log in again?
Folding Magnifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68112
100491-436EA 41.09 USD InStock
100491-436
Folding Magnifier, Electron Microscopy Sciences
Magnifiers
Lightweight, compact, and simple; it comes with a measuring scale on the base frame.
Base size is 1 square inch opening; marked every 1/8" on two sides and every millimeter on two other sides. Comes with 6X magnifier lens in a black anodized aluminum body. Folds flat when it is not in use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty