Would you like to log in again?
Light Magnifiers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68123 68123-L 68120 68121 68122
100491-440EA 89.39 USD InStock
100491-440 100492-718 100491-444 100491-442 100491-446
Light Magnifiers, Electron Microscopy Sciences
Magnifiers
3.5X Lighted Magnifier comes with a large square magnifier lens which measures 50 mm/side
A flashlight is attached to the magnifier that illuminates poorly lighted background making it bright and clear for easy observation. A variety of types and magnifications are available.
3.5X Lighted Magnifier comes with a large square magnifier lens which measures 50 mm/side. The lens is removable and the flashlight can be used alone.
7X and 10X Lighted Magnifiers are simple and accurate comparator. High quality measuring scale; detachable flashlight.
15X Lighted Magnifier is a high quality magnifier lens. Very convenient to see details under poor lighting conditions.
15X Lighted Scale Magnifier is a scale magnifier 15X is attached to an illuminating device to improve the visibility. Comes with one metric scale. Battery is not included.
The lens is removable and the flashlight can be used alone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty