Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68171 68181 68179 68173 68163 68182 68174 68162 68169 68164 68167 68178 68166 68184 68165 68170 68175 68161 68168 68172 68183 68180 68177 68176
100491-508EA 26.3 USD InStock
100491-508 100491-518 100491-506 100491-516 100491-478 100491-500 100491-522 100491-488 100491-510 100491-498 100491-520 100491-486 100491-504 100491-514 100491-502 100491-524 100491-512 100491-492 100491-480 100491-490 100491-496 100491-484 100491-494 100491-482
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Graticules Reticles and Grids
These scales are available in two different sizes, for use with the measuring magnifiers.
Available in 26 mm for 7× and some for 15× magnifiers and 35 mm for 10× magnifiers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty