Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Flat Embedding Molds, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70900 70902 70875 70870 70901 70903
100491-672EA 23.95 USD InStock
100491-672 100491-670 100491-676 100491-674 100491-668 100491-678
Flat Embedding Molds, Electron Microscopy Sciences
Specimen Mounting Supplies
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath.
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty