Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Microscope Slide Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71300
100491-770EA 75.03 USD InStock
100491-770
Microscope Slide Dispenser, Electron Microscopy Sciences
Slide Holders
Molded of attractive gray ABA plastic.
This dispenser provides a method of releasing clean, dust free 1 x 3" slides, one at a time. Comes with a clear molded plastic cover to allow for visual checking of unused slides. Holds approximately 72 slides.
Measurements: 4 3/8 x 4 7/8 x 5 1/2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty