Would you like to log in again?
Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71473 71472
100496-330EA 128.56 USD InStock
100496-330 100491-784
Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
Microscope Slide Boxes
PLEASE NOTE: For file up to 1,200 1x3" slides, refer to .
Molded one-piece polypropylene construction with an integral living hinge. Numbering in the base corresponds to the printed index in the covers for slide identification purpose. Boxes are designed to be stackable to save space. The tensioned clasp insures a secure closure. Available in 25 or 100 capacity. Fits 1 x 3" microscope slides.
PLEASE NOTE: For file up to 1,200 1x3" slides, refer to .
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty