Would you like to log in again?
Metal Polish, Pinky, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Pinky
71830
100491-816EA 16.35 USD InStock
100491-816
Metal Polish, Pinky, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Contains no silicone.
  • Good for cleaning EM parts
  • Blue paste
Grit Distribution: Particles over 45 µm diameter = 0 to 5%; 45 to 10 µm diameter = 20 to 40%; 10 to 1.6 µm diameter = 10 to 30%; under 1.6 µm diam = 40-60%.
* Vook, R.W., Kothari, R. (1992). Wear, 157, 65-79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty