Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Pen Vac™ Tool, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71914 71915
100501-340EA 152.3 USD InStock
100501-340 100491-856
Pen Vac™ Tool, Electron Microscopy Sciences
Vacuum Handling Systems
Pen-Vac™ is a new improved way to handle small, flat surface objects
  • Brushed aluminium body
  • Lifts upto 50 grams
  • Holds up to one minute
Pen-Vac™ comes with a variety of vacuum probes, complete with a vacuum cup attached. It is available with plastic or aluminum hubs and available in both straight and angled form. The stainless steel needle portion of the probes are one-half inch long. Vacuum Cups come in a wide range of materials. We offer the Static Dissipative and the Conductive Cups that provide ESD protection for electrostatic discharge of sensitive components. Cups comes in three sizes: 1/8" (3.17 mm); 1/4" (6.35 mm) and 3/8" (9.52 mm).
Beside the electronics assembly industry, jewelers, model builders etc. Pen-Vac is ideal for EM work as well. It can be used to handle grids, pick up stubs, align membranes, work with glass slides, cover slips and much more.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty