Would you like to log in again?
Coverglass Staining Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72240
100491-942EA 310.56 USD InStock
100491-942
Coverglass Staining Rack, Electron Microscopy Sciences
Staining Boxes
Coverglass staining rack is made from polished stainless steel.
  • Holds five of 22×24 mm coverglasses
  • Comes with a handle, 12.7 mm (5") long
This rack is specially designed to be used with coplin jars.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty